Merck
  • Surface chemical conversion of 3-glycidoxypropyldimethylethoxysilane on hydroxylated silicon surface: FT-IR, contact angle and ellipsometry analysis.
Surface chemical conversion of 3-glycidoxypropyldimethylethoxysilane on hydroxylated silicon surface: FT-IR, contact angle and ellipsometry analysis.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-10-16)
Serkan Demirci, Selin Kinali-Demirci, Tuncer Caykara
ABSTRACT
The chemical conversion of the top surface of 3-glycidoxypropyldimethylethoxysilane (GPDMES) on hyroxylated silicon surface has been studied as a function of reaction time. A multiple-step procedure was applied in this study. At first, GPDMES molecules were self-assembled on the hydroxylated silicon surface. The second step was the modification of epoxy groups with 3,3'-iminodipropionitrile and the last step was the amidoximation reaction of nitrile groups. Existence of the monolayers covalently attached to silicon surfaces were revealed by X-ray photoelectron spectroscopy and Fourier transform infrared spectroscopy (FT-IR). Modification, conversion and thickness of surfaces were followed by FT-IR and ellipsometry analysis.
Sigma-Aldrich
3,3′-Iminodipropionitrile, technical grade, 90%