Merck
  • Synthesis and antiviral activities of hexadecyloxypropyl prodrugs of acyclic nucleoside phosphonates containing guanine or hypoxanthine and a (S)-HPMP or PEE acyclic moiety.
Synthesis and antiviral activities of hexadecyloxypropyl prodrugs of acyclic nucleoside phosphonates containing guanine or hypoxanthine and a (S)-HPMP or PEE acyclic moiety.
European journal of medicinal chemistry (2012-08-04)
Tomáš Tichý, Graciela Andrei, Robert Snoeck, Jan Balzarini, Martin Dračínský, Marcela Krečmerová
ABSTRACT
Hexadecyloxypropyl esters of acyclic nucleoside phosphonates containing guanine (G) or hypoxanthine (Hx) and a (S)-[3-hydroxy-2-(phosphonomethoxy)propyl] [(S)-HPMP] or 2-(2-phosphonoethoxy)ethyl (PEE) acyclic moiety have been prepared. The activity of the prodrugs was evaluated in vitro against different virus families. Whereas ester derivatives of PEEHx and (S)-HPMPHx were antivirally inactive, monoesters of PEEG, and mono- and diesters of (S)-HPMPG showed pronounced antiviral activity against vaccinia virus and/or herpesviruses. Monoesters of (S)-HPMPG emerged as the most potent and selective derivatives against these DNA viruses. None of the compounds were inhibitory against RNA viruses and retroviruses.
Sigma-Aldrich
Hypoxanthine, Vetec, reagent grade, 98%
Sigma-Aldrich
Hypoxanthine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Hypoxanthine, ≥99.0%