Merck
  • Quantification of the histochemical staining for carbonyles and DNA using 3-hydroxy-2-naphthoic acid hydrazide and fast blue B.
Quantification of the histochemical staining for carbonyles and DNA using 3-hydroxy-2-naphthoic acid hydrazide and fast blue B.
Histochemistry (1990-01-01)
G Nöhammer
ABSTRACT
Fixed cells and tissues pretreated with 4-hydroxynonenal were used as models for the histochemical demonstration of protein bound aldehydic groups. The aldehydes were stained with both a modification of the 2,4-dinitrophenylhydrazine method (2,4-DNPH) and the optimized staining using 3-hydroxy-2-naphthoic acid hydrazide and Fast blue B (NAH-FB). A correlation has been found between the specific microphotometric mean integrated maximum absorbance values of cells and tissues stained with 2,4-DNPH and with NAH-FB (cc = 0.999). The maximum absorbance measured after 2,4-DNPH-staining (epsilon 367 = 21,000) were 1.893 +/- 0.072 (P less than 0.01) times that of NAH-FB-staining at 550 nm. Microphotometrically determined DNA-values of different cells stained with the NAH-FB-DNA-method correlated with those determined with methods of analytical biochemistry and published by other authors.
Sigma-Aldrich
Fast Blue B Salt, Dye content ~95 %