Merck
  • Oxidation-mediated DNA cross-linking contributes to the toxicity of 6-thioguanine in human cells.
Oxidation-mediated DNA cross-linking contributes to the toxicity of 6-thioguanine in human cells.
Cancer research (2012-07-24)
Reto Brem, Peter Karran
ABSTRACT
The thiopurines azathioprine and 6-mercaptopurine have been extensively prescribed as immunosuppressant and anticancer agents for several decades. A third member of the thiopurine family, 6-thioguanine (6-TG), has been used less widely. Although known to be partly dependent on DNA mismatch repair (MMR), the cytotoxicity of 6-TG remains incompletely understood. Here, we describe a novel MMR-independent pathway of 6-TG toxicity. Cell killing depended on two properties of 6-TG: its incorporation into DNA and its ability to act as a source of reactive oxygen species (ROS). ROS targeted DNA 6-TG to generate potentially lethal replication-arresting DNA lesions including interstrand cross-links. These triggered processing by the Fanconi anemia and homologous recombination DNA repair pathways. Allopurinol protected against 6-TG toxicity by acting as a ROS scavenger and preventing DNA damage. Together, our findings provide mechanistic evidence to support the proposed use of thiopurines to treat HR-defective tumors and for the coadministration of 6-TG and allopurinol as an immunomodulation strategy in inflammatory disorders.
Sigma-Aldrich
6-Thioguanine, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
6-Thioguanine, Hybri-Max, 50 ×, γ-irradiated, lyophilized powder, BioXtra, suitable for hybridoma
Pricing and availability is not currently available.