Merck
  • Cefoxitin as an alternative to carbapenems in a murine model of urinary tract infection due to Escherichia coli harboring CTX-M-15-type extended-spectrum β-lactamase.
Cefoxitin as an alternative to carbapenems in a murine model of urinary tract infection due to Escherichia coli harboring CTX-M-15-type extended-spectrum β-lactamase.
Antimicrobial agents and chemotherapy (2012-01-05)
Raphaël Lepeule, Etienne Ruppé, Patrick Le, Laurent Massias, Françoise Chau, Amandine Nucci, Agnès Lefort, Bruno Fantin
ABSTRACT
We investigated the efficiency of the cephamycin cefoxitin as an alternative to carbapenems for the treatment of urinary tract infections (UTIs) due to Escherichia coli producing CTX-M-type extended-spectrum β-lactamases. The susceptible, UTI-inducing E. coli CFT073-RR strain and its transconjugant CFT073-RR Tc (pbla(CTX-M-15)), harboring a bla(CTX-M-15) carrying-plasmid, were used for all experiments. MICs of cefoxitin (FOX), ceftriaxone (CRO), imipenem (IMP), and ertapenem (ETP) for CFT073-RR and CFT073-RR Tc (pbla(CTX-M-15)) were 4 and 4, 0.125 and 512, 0.5 and 0.5, and 0.016 and 0.032 μg/ml, respectively. Bactericidal activity was similarly achieved in vitro against the two strains after 3 h of exposure to concentrations of FOX, IMI, and ETP that were 2 times the MIC, whereas CRO was not bactericidal against CFT073-RR Tc (pbla(CTX-M-15)). The frequencies of spontaneous mutants of the 2 strains were not higher for FOX than for IMP or ETP. In the murine model of UTIs, mice infected for 5 days were treated over 24 h. Therapeutic regimens in mice (200 mg/kg of body weight every 3 h or 4 h for FOX, 70 mg/kg every 6 h for CRO, 100 mg/kg every 2 h for IMP, and 100 mg/kg every 4 h for ETP) were chosen in order to reproduce the percentage of time that free-drug concentrations above the MIC are obtained in humans with standard regimens. All antibiotic regimens produced a significant reduction in bacterial counts (greater than 2 log(10) CFU) in kidneys and bladders for both strains (P < 0.001) without selecting resistant mutants in vivo, but the reduction obtained with CRO against CFT073-RR Tc (pbla(CTX-M-15)) in kidneys was significantly lower than that obtained with FOX. In conclusion, FOX appears to be an effective therapeutic alternative to carbapenems for the treatment of UTIs due to CTX-M-producing E. coli.
Product Description
Cefoxitin sodium, European Pharmacopoeia (EP) Reference Standard