Merck
  • Nickel-catalyzed Heck-type reactions of benzyl chlorides and simple olefins.
Nickel-catalyzed Heck-type reactions of benzyl chlorides and simple olefins.
Journal of the American Chemical Society (2011-11-10)
Ryosuke Matsubara, Alicia C Gutierrez, Timothy F Jamison
ABSTRACT
Nickel-catalyzed intermolecular benzylation and heterobenzylation of unactivated alkenes to provide functionalized allylbenzene derivatives are described. A wide range of both the benzyl chloride and alkene coupling partners are tolerated. In contrast to analogous palladium-catalyzed variants of this process, all reactions described herein employ electronically unbiased aliphatic olefins (including ethylene), proceed at room temperature, and provide 1,1-disubstituted olefins over the more commonly observed 1,2-disubstituted olefins with very high selectivity.
Sigma-Aldrich
Benzyl chloride, ReagentPlus®, 99%, contains ≤1% propylene oxide as stabilizer
Sigma-Aldrich
Allylbenzene, 98%
Sigma-Aldrich
Benzyl chloride, AR, ≥99%