Merck
  • Oncostatin M is a major mediator of cardiomyocyte dedifferentiation and remodeling.
Oncostatin M is a major mediator of cardiomyocyte dedifferentiation and remodeling.
Cell stem cell (2011-11-08)
Thomas Kubin, Jochen Pöling, Sawa Kostin, Praveen Gajawada, Stefan Hein, Wolfgang Rees, Astrid Wietelmann, Minoru Tanaka, Holger Lörchner, Silvia Schimanski, Marten Szibor, Henning Warnecke, Thomas Braun
ABSTRACT
Cardiomyocyte remodeling, which includes partial dedifferentiation of cardiomyocytes, is a process that occurs during both acute and chronic disease processes. Here, we demonstrate that oncostatin M (OSM) is a major mediator of cardiomyocyte dedifferentiation and remodeling during acute myocardial infarction (MI) and in chronic dilated cardiomyopathy (DCM). Patients suffering from DCM show a strong and lasting increase of OSM expression and signaling. OSM treatment induces dedifferentiation of cardiomyocytes and upregulation of stem cell markers and improves cardiac function after MI. Conversely, inhibition of OSM signaling suppresses cardiomyocyte remodeling after MI and in a mouse model of DCM, resulting in deterioration of heart function after MI but improvement of cardiac performance in DCM. We postulate that dedifferentiation of cardiomyocytes initially protects stressed hearts but fails to support cardiac structure and function upon continued activation. Manipulation of OSM signaling provides a means to control the differentiation state of cardiomyocytes and cellular plasticity.
Sigma-Aldrich
Oncostatin M from mouse, BioReagent, ≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Oncostatin M human, OSM, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture