Merck
  • A protocol to 2-aminobenzimidazoles via copper-catalyzed cascade addition and cyclization of o-haloanilines and carbodiimides.
A protocol to 2-aminobenzimidazoles via copper-catalyzed cascade addition and cyclization of o-haloanilines and carbodiimides.
The Journal of organic chemistry (2011-03-19)
Fei Wang, Shangjun Cai, Qian Liao, Chanjuan Xi
ABSTRACT
An efficient strategy for the synthesis of a variety of 2-animobenzimidazole derivatives has been developed. The reaction proceeded from o-haloanilines and carbodiimides via copper(I)-catalyzed domino reaction in the presence of tert-butoxide to afford the corresponding 2-animobenzimidazole derivatives in good to excellent yields. o-Haloanilines could be o-iodoaniline, o-bromoaniline, and o-chloroaniline derivatives. Carbodiimides could be symmetrical and unsymmetrical substrates with aryl or alkyl substituents. The reaction exhibited a good regioselectivity when unsymmetrical carbodiimides were employed.
Sigma-Aldrich
2-Aminobenzimidazole, 97%