Merck
  • Antioxidative activity of microbial metabolites of (-)-epigallocatechin gallate produced in rat intestines.
Antioxidative activity of microbial metabolites of (-)-epigallocatechin gallate produced in rat intestines.
Bioscience, biotechnology, and biochemistry (2011-03-11)
Akiko Takagaki, Shuichi Otani, Fumio Nanjo
ABSTRACT
The antioxidative activities of (-)-epigallocatechin gallate (EGCg) metabolites degraded by rat intestinal flora were investigated by a flow injection analysis coupled to an on-line antioxidant detection system with the 2,2'-azinobis (3-ethylbenzothiazoline-6-sulfonic acid) (ABTS) radical cation. All of the metabolites were found to have antioxidative activity, suggesting that the EGCg metabolites may show antioxidative activity in the body.
Sigma-Aldrich
γ-Valerolactone, natural, ≥95%, FG
Sigma-Aldrich
γ-Valerolactone, ReagentPlus®, 99%
Sigma-Aldrich
γ-Valerolactone, ≥99%, FCC, FG