Merck
  • Pharmacokinetics and biodistribution of lonidamine/paclitaxel loaded, EGFR-targeted nanoparticles in an orthotopic animal model of multi-drug resistant breast cancer.
Pharmacokinetics and biodistribution of lonidamine/paclitaxel loaded, EGFR-targeted nanoparticles in an orthotopic animal model of multi-drug resistant breast cancer.
Nanomedicine : nanotechnology, biology, and medicine (2011-01-12)
Lara Milane, Zhen-feng Duan, Mansoor Amiji
ABSTRACT
The aim of this study was to assess the biodistribution and pharmacokinetics of epidermal growth factor receptor (EGFR)-targeted polymer-blend nanoparticles loaded with the anticancer drugs lonidamine and paclitaxel. Plasma, tumor, and tissue distribution profiles were quantified in an orthotopic animal model of multidrug-resistant breast cancer and were compared to treatment with nontargeted nanoparticles and to treatment with drug solution. A poly(d,l-lactide-co-glycolide)-poly(ethylene glycol)-EGFR targeting peptide (PLGA-PEG-EFGR peptide) construct was synthesized for incorporation in poly(ɛ-caprolactone) particles to achieve active EGFR targeting. An isocratic high-pressure liquid chromatography method was developed to quantify lonidamine and paclitaxel in mice plasma, tumors, and vital organs. The targeted nanoparticles demonstrated a superior pharmacokinetic profile relative to drug solution and nontargeted nanoparticles, particularly for lonidamine delivery. The first target site of accumulation was the liver, followed by the kidneys, and then the tumor mass; maximal tumor accumulation occured at 3 hours after administration. Lonidamine-paclitaxel combination therapy administered via EGFR-targeted polymer-blend nanocarriers may become a viable platform for the future treatment of multidrug-resistant cancer. In this study the biodistribution and pharmacokinetics of epidermal growth factor receptor (EGFR)-targeted polymer-blend nanoparticles loaded with lonidamine and paclitaxel were assessed. The targeted nanoparticles demonstrated a superior pharmacokinetic profile relative to drug solution and nontargeted nanoparticles, paving the way to new therapeutic approaches for multidrug-resistant malignancies.