Merck
  • In vitro conversion of model sulfenamide prodrugs in the presence of small molecule thiols.
In vitro conversion of model sulfenamide prodrugs in the presence of small molecule thiols.
Journal of pharmaceutical sciences (2010-10-20)
Kwame W Nti-Addae, Valentino J Stella
ABSTRACT
Sulfenamide prodrugs of amide and urea functional group containing drugs have recently been proposed as a means of altering the physical and bioproperties of problematic drug molecules containing these two functionalities. Sulfenamides have been shown to revert to the parent drug via reaction with thiols. Explored here is the mechanism for this reaction. The stoichiometry and pH dependency of the in vitro reversion of two model prodrugs of the oxazolidinone, linezolid, and a sulfenamide of phthalimide were studied at 25 °C in the presence of thiols, including cysteine and glutathione, of varying basicity. High-performance/pressure liquid chromatography and liquid chromatography-mass spectrometry results showed the near quantitative reversion of the sulfenamides to the parent drug with simultaneous formation of a mixed disulfide. The pH and the dependency of the reaction on the basicity of the thiol strongly supported the role of the thiolate species in the conversion. The reaction is consistent with an S(N)2 type mechanism seen in the reaction of some thiols with disulfides.
Sigma-Aldrich
Sulfamerazine, ReagentPlus®, ≥99.0%
Supelco
Sulfamerazine, VETRANAL®, analytical standard