Merck
  • Carboxylic acid appended dipyrrin for the formation of a hexanuclear iridium/copper paddlewheel complex.
Carboxylic acid appended dipyrrin for the formation of a hexanuclear iridium/copper paddlewheel complex.
Inorganic chemistry (2010-09-02)
Catherine Bronner, Stéphane A Baudron, Mir Wais Hosseini
ABSTRACT
Reaction of a heteroleptic cyclometalated iridium complex incorporating a carboxylic acid appended dipyrrin with copper acetate leads to the formation of a hexanuclear species with the paddlewheel structure.
Sigma-Aldrich
Copper(II) acetate, powder, 99.99% trace metals basis
Sigma-Aldrich
Copper(II) acetate, LR, ≥98%
Sigma-Aldrich
Copper(II) acetate, 98%