Merck
  • Transformation of arylamines into direct-acting mutagens by reaction with nitrite.
Transformation of arylamines into direct-acting mutagens by reaction with nitrite.
Mutation research (1991-07-01)
T Kato, N Tadokoro, M Tsutsui, K Kikugawa
ABSTRACT
Arylamines including aniline (I), 1-naphthylamine (II), 2-naphthylamine (III), 2-aminofluorene (IV), 1-aminoanthracene (V) and 1-aminopyrene (VI) were treated with 4 equivalent amounts of nitrite at pH3 and 37 degrees C for 4 h. The reaction mixtures of I, IV, V and VI showed mutagenicity to Salmonella typhimurium TA98 and TA100 strains without metabolic activation. The numbers of His+ revertant colonies to TA98 strain were 110/0.05 mumole I, 970/0.055 mumole IV, 620/0.10 mumole V and 870/0.02 mumole VI. These arylamines were converted into mutagens with diazoquinone, diazonium and nitro functions depending on their structures. The mutagen from I was p-diazoquinone (I2). The mutagen from IV was highly unstable fluorene-2-diazonium salt (IV1). The mutagens from V were N3O3-introduced anthracene (V1-1) and 1-nitroanthracene (V2), and those from VI were unidentified nitro-introduced compound (VI1) and 1-nitropyrene (VI2).
Sigma-Aldrich
1-Aminoanthracene, technical grade, 90%