Merck
  • Study of the effect of Wuzhi tablet (Schisandra sphenanthera extract) on tacrolimus tissue distribution in rat by liquid chromatography tandem mass spectrometry method.
Study of the effect of Wuzhi tablet (Schisandra sphenanthera extract) on tacrolimus tissue distribution in rat by liquid chromatography tandem mass spectrometry method.
Biomedical chromatography : BMC (2009-08-21)
Xiao-ling Qin, Hui-chang Bi, Chang-xi Wang, Jia-li Li, Xue-ding Wang, Long-shan Liu, Xiao Chen, Min Huang
ABSTRACT
A liquid chromatography/tandem mass spectrometry (LC-MS/MS) method was developed and validated for determining tacrolimus (FK506) in rat tissues to study the effect of Schisandra sphenanthera extract on FK506 tissue distribution. After a liquid-liquid extraction with ethyl acetate, FK506 and ascomycin (IS) were subjected to LC-MS/MS analysis using positive electrospray ionization under multiple reactions monitoring mode. Chromatographic separation of FK506 and ascomycin was achieved on a Hypersil BDS C(18) column with a mobile phase consisting of methanol-water (containing 2 mM ammonium acetate, 95 : 5, v/v). The intra- and inter-batch precision of the method were less than 8.8 and 9.8%, respectively. The intra- and inter-batch accuracies ranged from 97.5 to 104.0%. The lowest limit of quantification for FK506 was 0.5 ng/mL. The method was applied to a FK506 tissue distribution study with or without a dose of Wuzhi (WZ) tablet. Most of the FK506 tissue concentrations were slightly increased after a concomitant WZ tablet dose, but the whole blood concentration of FK506 was dramatically increased 3-fold after a concomitant WZ tablet dose. These results indicated that the LC-MS/MS method was rapid and sensitive enough to quantify FK506 in different rat tissues, and strict drug monitoring is recommended when co-administering WZ tablet in clinical use.