Merck
  • Probenecid, an inhibitor of transmembrane organic anion transporters, alters tissue distribution of DNA adducts in 1-hydroxymethylpyrene-treated rats.
Probenecid, an inhibitor of transmembrane organic anion transporters, alters tissue distribution of DNA adducts in 1-hydroxymethylpyrene-treated rats.
Toxicology (2009-06-02)
Bernhard H Monien, Carolin Müller, Nadiya Bakhiya, Claudia Donath, Heinz Frank, Albrecht Seidel, Hansruedi Glatt
ABSTRACT
1-Methylpyrene (1-MP), an abundant alkylated polycyclic aromatic hydrocarbon, is activated by side-chain hydroxylation to 1-hydroxymethylpyrene (1-HMP) and subsequent sulfo-conjugation to electrophilic 1-sulfooxymethylpyrene (1-SMP). In rats, this activation mainly occurs in liver. 1-SMP may react with hepatic DNA or be exported into the blood circulation to reach other tissues, in particular kidneys. Findings with recombinant cell lines suggest that renal 1-SMP uptake proceeds via organic anion transporters (OATs). Here, we tested the hypothesis that probenecid, a characteristic OAT inhibitor, interferes with kidney damage brought about by 1-SMP formed in rats. 1-HMP was administered intraperitoneally to 30 rats, half of which were co-treated with probenecid. The tissue distribution of DNA adducts was analyzed using (32)P-postlabeling and isotope dilution LC-MS/MS for the detection of the adducts N(2)-(1-methylpyrenyl)-2'-deoxyguanosine and N(6)-(1-methylpyrenyl)-2'-deoxyadenosine. In rats treated solely with 1-HMP, adduct levels in kidney tissue were about 3-fold and 8-fold higher than those in liver and lung, respectively. After co-treatment with probenecid, hepatic and pulmonary adduct levels were 12-fold and 4-fold elevated, respectively, whereas renal adduct levels were slightly lower compared to those of rats receiving 1-HMP alone. Moreover, serum levels of 1-SMP were increased 23-fold in animals pre-treated with probenecid. The differential effects on hepatic and pulmonary adduct levels suggest that not only renal OATs, but also additional anion transporters, e.g. those mediating the hepatic export of 1-SMP into the bile, were inhibited. Thus, transmembrane transport proteins play a crucial role in the distribution of reactive phase II metabolites, and thereby in tissue allocation of DNA adducts.
Sigma-Aldrich
1-Pyrenemethanol, 98%