- Synthesis of functionalised diarylmethanes via a cobalt-catalysed cross-coupling of arylzinc species with benzyl chlorides.
Synthesis of functionalised diarylmethanes via a cobalt-catalysed cross-coupling of arylzinc species with benzyl chlorides.
Chemical communications (Cambridge, England) (2008-10-22)
Muriel Amatore, Corinne Gosmini
PMID18931772
ABSTRACT
A new cobalt-catalysed reductive coupling of aryl halides with benzyl chlorides is reported; a variety of diarylmethanes can be prepared in good to excellent yields under mild reaction conditions using CoBr(2) as catalyst and Zn dust; this new cobalt-catalysed coupling represents a practical and interesting alternative to previously known methods for the synthesis diarylmethanes.