Merck
  • Chemical and optical control of peristaltic actuator based on self-oscillating porous gel.
Chemical and optical control of peristaltic actuator based on self-oscillating porous gel.
Chemical communications (Cambridge, England) (2008-10-03)
Shu-Ichi Shinohara, Takahiro Seki, Takamasa Sakai, Ryo Yoshida, Yukikazu Takeoka
ABSTRACT
We demonstrate the chemical and optical control of the self-sustaining peristaltic motion of a structural colored porous hydrogel.
Product Description
Supelco
Sodium bromate, puriss. p.a., ≥99.5% (RT)
Sigma-Aldrich
Sodium bromate, ≥99%