Merck
Aqueous solutions of transition metal containing micelles.
Advances in colloid and interface science (2008-09-25)
Peter C Griffiths, Ian A Fallis, Thomas Tatchell, Lisa Bushby, Andrew Beeby
ABSTRACT
Incorporation of d- or f-block metals into ligand systems that renders a metal complex surface-active or drives its partitioning into surfactant phases enables the localisation of chemical functionality at interfaces. This article discusses a number of fundamental aspects of these interesting materials and examines potential applications.
Sigma-Aldrich
Bis(4-nitrophenyl) phosphate, 99%
Sigma-Aldrich
Bis(p-nitrophenyl) phosphate sodium salt, ≥99% (TLC)