- Fragrance material review on geranodyle.
Fragrance material review on geranodyle.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2008-07-22)
S P Bhatia, D McGinty, C S Letizia, A M Api
PMID18640183
ABSTRACT
A toxicologic and dermatologic review of geranodyle when used as a fragrance ingredient is presented.