Merck
  • Assay of fatty acid omega-hydroxylation using high-performance liquid chromatography with fluorescence labeling reagent, 3-bromomethyl-7-methoxy-1,4-benzoxazin-2-one (BrMB).
Assay of fatty acid omega-hydroxylation using high-performance liquid chromatography with fluorescence labeling reagent, 3-bromomethyl-7-methoxy-1,4-benzoxazin-2-one (BrMB).
Analytical biochemistry (1991-11-15)
J Yamada, M Sakuma, T Suga
ABSTRACT
A sensitive nonradioisotopic method is reported for measuring microsomal lauric acid omega-hydroxylation activity. The assay is based upon separation and detection of 12-hydroxylauric acid formed by means of high-performance liquid chromatography following fluorescence labeling of the carboxyl group with 3-bromomethyl-7-methoxy-1,4-benzoxazin-2-one (BrMB). The use of 10-hydroxycapric acid as an internal standard affords the accurate and reproducible assay. The differential effect of dehydroepiandrosterone, a peroxisome proliferator, on the omega-hydroxylation activity in the liver and kidney of rats is also reported.