Merck
  • Using silica particles to isolate total RNA from plant tissues recalcitrant to extraction in guanidine thiocyanate.
Using silica particles to isolate total RNA from plant tissues recalcitrant to extraction in guanidine thiocyanate.
Analytical biochemistry (2008-01-02)
Ling-Wen Ding, Qiao-Yang Sun, Zhao-Yu Wang, Yong-Bin Sun, Zeng-Fu Xu
ABSTRACT
The most commonly used protocol of the RNA isolation, the guanidine thiocyanate method, was unsuitable for recalcitrant plant tissues containing a large amount of storage proteins and secondary metabolites. We demonstrated that RNA could bind to the silica particles, which have been used successfully in DNA isolation from various sources, under a high concentration of NaCl in the presence of ethanol and sodium acetate. Based on this observation, an efficient, inexpensive, and highly reproducible technique, the acid phenol-silica method, was developed to isolate high-quality RNAs from various plant tissues recalcitrant to extraction in guanidine thiocyanate.
Sigma-Aldrich
Guanidine thiocyanate, BioUltra, for molecular biology, ≥99.0% (AT)
Sigma-Aldrich
Guanidine thiocyanate, BioReagent, for molecular biology, ≥99%
Sigma-Aldrich
Guanidine thiocyanate solution, BioUltra, for molecular biology, ~6 M in H2O
Sigma-Aldrich
Guanidine thiocyanate, for molecular biology, ≥99%
Sigma-Aldrich
Guanidine thiocyanate, ≥97% (titration)
Sigma-Aldrich
Guanidine thiocyanate, Vetec, reagent grade, ≥97%