Merck
  • Cholinesterase activities as potential biomarkers: characterization in two freshwater snails, Potamopyrgus antipodarum (Mollusca, Hydrobiidae, Smith 1889) and Valvata piscinalis (Mollusca, Valvatidae, Müller 1774).
Cholinesterase activities as potential biomarkers: characterization in two freshwater snails, Potamopyrgus antipodarum (Mollusca, Hydrobiidae, Smith 1889) and Valvata piscinalis (Mollusca, Valvatidae, Müller 1774).
Chemosphere (2007-11-14)
Beatrice Gagnaire, Olivier Geffard, Benoit Xuereb, Christelle Margoum, Jeanne Garric
ABSTRACT
Anti-cholinesterase insecticides constitute a major portion of modern synthetic pesticides and the assessment of cholinesterase (ChE) inhibition is widely used as a specific biomarker for evaluating the exposure of non-target organisms to these pollutants. However, most studies on this biomarker were developed on vertebrates and among invertebrates, gastropod mollusks are rarely used. Gastropods are important members of aquatic habitats and therefore present a high ecological relevance for freshwater ecosystems. In this context, ChE activities were characterized in two freshwater gastropod mollusks, Potamopyrgus antipodarum and Valvata piscinalis, in order to ascertain their value as sentinel species. Firstly, characterization of ChE activities was performed using different substrates (acetylcholine iodide, butyrylcholine iodide and propionylcholine iodide) and specific inhibitors (eserine, iso-OMPA and BW284c51). Secondly, in vivo effect of a widely used organophosphate insecticide, chlorpyrifos, was tested on ChE activity in both species. Results suggested that P. antipodarum possesses two isoforms of cholinesterases, one isoform which properties are intermediate between an acetyl and a propionyl ChE, and one minor isoform which correspond to a butyryl ChE, while V. piscinalis seems to possess only one isoform which displays typical properties of an acetyl ChE. Chlorpyrifos induced no effect on V. piscinalis ChE. In contrast, P. antipodarum activity was significantly decreased by environmental realistic chlorpyrifos concentrations (2.86 and 14.2 nM) after seven days of contact. The present study suggests that P. antipodarum may be employed as a biological indicator for assessing pesticide contamination.
Sigma-Aldrich
Propionylthiocholine iodide, ≥98%