Merck
  • Selective oxidation of methanol to dimethoxymethane under mild conditions over V2O5/TiO2 with enhanced surface acidity.
Selective oxidation of methanol to dimethoxymethane under mild conditions over V2O5/TiO2 with enhanced surface acidity.
Chemical communications (Cambridge, England) (2007-05-24)
Yuchuan Fu, Jianyi Shen
ABSTRACT
Dimethoxymethane was synthesized from the direct oxidation of methanol with high conversion and selectivity over specially designed bifunctional V(2)O(5)/TiO2 catalysts with redox and enhanced acidic character, in which the surface acidity played an essential role for inhibiting the formation of formaldehyde through the enhanced condensation reaction of formaldehyde with methanol to produce dimethoxymethane.
Sigma-Aldrich
Dimethoxymethane, ReagentPlus®, 99%
Sigma-Aldrich
Formaldehyde dimethyl acetal, absolute, over molecular sieve (H2O ≤0.01%), ≥99.0% (GC)
Sigma-Aldrich
Formaldehyde dimethyl acetal, for Grignard reactions, ≥99.0% (GC)