Merck
  • Glow discharge plasma treatment of polyethylene tubing with tetraglyme results in ultralow fibrinogen adsorption and greatly reduced platelet adhesion.
Glow discharge plasma treatment of polyethylene tubing with tetraglyme results in ultralow fibrinogen adsorption and greatly reduced platelet adhesion.
Journal of biomedical materials research. Part A (2006-08-03)
Lan Cao, Sivaprasad Sukavaneshvar, Buddy D Ratner, Thomas A Horbett
ABSTRACT
Previous studies from our lab have shown that fibrinogen adsorption (Gamma(Fg)) must be reduced below 10 ng/cm(2) to significantly reduce platelet adhesion, and that radio frequency glow discharge (RFGD) treatment of polymeric films in the presence of tetraethylene glycol dimethyl ether (tetraglyme) can reduce Gamma(Fg) to the desired ultralow value. In this report, the effects of RFGD coatings of tetraglyme on the lumenal surface of PE tubing on Gamma(Fg) and on blood interactions both in vitro and ex vivo are described. Gamma(Fg) on the tetraglyme-coated PE tubing was reduced to the desired ultralow level (<10 ng/cm(2)), and we also observed a significant decrease in adsorption of von Willebrand's factor. In vitro platelet adhesion from washed platelet suspensions, platelet rich plasma, or whole blood to tetraglyme-coated PE tubing was decreased compared to PE, polyurethane, or silicone rubber tubes. In addition, thrombin generation by platelets adherent to tetraglyme-coated PE was also much less than by platelets adherent to PE. When inserted in an ex vivo carotid artery-carotid artery shunt in sheep, the RFGD tetraglyme-coated PE exhibited a very low number of adherent platelets compared to heparin-coated, chromic acid-etched, or plain PE. The RFGD tetraglyme-coated PE tubes exhibited high protein and platelet resistance in vitro, and high platelet resistance ex vivo. The improved hemocompatibility is attributed to the unique chemical structure of RFGD tetraglyme that makes it highly protein resistant.
Sigma-Aldrich
Tetraethylene glycol dimethyl ether, ≥99%