Merck
  • Direct enantio- and diastereoselective Mannich reactions of malonate and beta-keto esters with N-Boc and N-Cbz aldimines catalysed by a bifunctional cinchonine derivative.
Direct enantio- and diastereoselective Mannich reactions of malonate and beta-keto esters with N-Boc and N-Cbz aldimines catalysed by a bifunctional cinchonine derivative.
Chemical communications (Cambridge, England) (2006-03-07)
A Louise Tillman, Jinxing Ye, Darren J Dixon
ABSTRACT
A highly enantioselective Mannich reaction between malonate esters and N-Boc and N-Cbz aldimines, catalysed by a bifunctional cinchonine derivative, has been developed; extension of this methodology to encompass the use of 2-substituted-1,3-dicarbonyl nucleophiles allows the formation of adjacent stereocentres, one of which is quaternary, in high relative and absolute stereocontrol.
Sigma-Aldrich
(+)-Cinchonine, 85%
Sigma-Aldrich
Cinchonine, crystallized, ≥98.0% (NT)