Merck
  • An alpha/beta-fold C--C bond hydrolase is involved in a central step of nicotine catabolism by Arthrobacter nicotinovorans.
An alpha/beta-fold C--C bond hydrolase is involved in a central step of nicotine catabolism by Arthrobacter nicotinovorans.
Journal of bacteriology (2005-12-03)
Paula Sachelaru, Emile Schiltz, Gabor L Igloi, Roderich Brandsch
ABSTRACT
The enzyme catalyzing the hydrolytic cleavage of 2,6-dihydroxypseudooxynicotine to 2,6-dihydroxypyridine and gamma-N-methylaminobutyrate was found to be encoded on pAO1 of Arthrobacter nicotinovorans. The new enzyme answers an old question about nicotine catabolism and may be the first C--C bond hydrolase that is involved in the biodegradation of a heterocyclic compound.
Sigma-Aldrich
2,6-Dihydroxypyridine hydrochloride, 97%