Merck
Metabolism of nilutamide in rat lung.
Biochemical pharmacology (2005-11-30)
Kjetil Ask, Nathalie Décologne, Christian Ginies, Marit Låg, Jean Luc Boucher, Jørn A Holme, Hélène Pelczar, Philippe Camus
ABSTRACT
Nilutamide is a non-steroidal anti-androgen drug proposed in the treatment of metastatic prostatic carcinoma. Its therapeutic effects are overshadowed by the occurrence of adverse reactions, mediated by mechanisms that remain elusive. To elucidate possible mechanisms for nilutamide toxicity, we investigated the metabolism of nilutamide in rat lung homogenates, in subcellular fractions and in freshly isolated cells. In whole lung homogenates, the nitro group of nilutamide was reduced to the amine and hydroxylamine moieties. These conversions occurred exclusively in the absence of dioxygen, were increased by the addition of FMN, FAD, or NADPH. Reductive metabolism of nilutamide to the amine and hydroxylamine was further evidenced in subcellular fractions obtained by differential ultracentrifugation. It was found to take place mainly in the cytosol of rat lung and to be stimulated, strongly, upon co-addition of NADPH and FMN. Addition of inhibitors of enzymes involved in the reductive metabolism of nitroaromatic compounds indicated that reduction of nilutamide involved, mainly, soluble flavoproteins. Incubations with freshly isolated lung cells revealed that macrophages were the main players in nitroreduction of nilutamide whereas the epithelial type II cells and the non-ciliated Clara cells were less efficient in catalyzing this reaction. Our results show that nilutamide is extensively reduced by lung tissues in the absence of oxygen, especially by enzymes found in alveolar macrophages. In accordance with recent findings, subcellular localization, oxygen sensitivity, cofactor requirements and inhibitor studies lead us to suggest the involvement of a soluble nitric oxide synthase in lung cytosolic nitroreduction.
Sigma-Aldrich
Nilutamide, solid