Merck
  • [Experiment study on the estrogen-like effect of compounds of mercury, chromium and manganese].
[Experiment study on the estrogen-like effect of compounds of mercury, chromium and manganese].
Wei sheng yan jiu = Journal of hygiene research (2005-05-03)
Ya-Dong Wang, Xiao-Yu Chen, Yi-Ming Wu, Dong Xu
ABSTRACT
In order to study estrogen-like effect of compounds of Mercury, Chromium and Manganese. Mercury Chloride, Manganese Sulfate, Chromium Chloride and Chromium trioxide were selected to perform proliferation assay of MCF-7 human breast cancer cells and binding assay of estrogen receptor for rat uterine. Mercury Chloride could stimulate the proliferation of MCF-7 cell, the maximal increase of MCF-7 cells was measured by Mercury Chloride at 10(-7) mol/L. The effect was blocked completely by a pure antigestrogen ICI182, 780; but Mercury Chloride could not bind to ER by competing with E2. Chromium Chloride, Chromium Trioxide and Manganese Sulfate were not able to stimulate the proliferation of MCF-7 cell and bind to ER through competing against 3H-E2 in vitro. Mercury Chloride exhibited the estrogenicity through binding and activating ER; Chromium Chloride, Chromium trioxide and Manganese Sulfate did not show any estrogenicity.
Sigma-Aldrich
Chromium(VI) oxide, 99.99% trace metals basis
Sigma-Aldrich
Chromium(VI) oxide, LR, ≥99%
Sigma-Aldrich
Chromium(VI) oxide, ACS reagent, ≥98.0%
Sigma-Aldrich
Chromium(VI) oxide, ReagentPlus®, 99.9% trace metals basis