Merck
  • Chloroperoxidase from Caldariomyces fumago is active in the presence of an ionic liquid as co-solvent.
Chloroperoxidase from Caldariomyces fumago is active in the presence of an ionic liquid as co-solvent.
Biotechnology letters (2005-01-27)
Claudia Sanfilippo, Nicola D'Antona, Giovanni Nicolosi
ABSTRACT
Chloroperoxidase from Caldariomyces fumago catalyses the oxidation of 1,2-dihydronaphthalene to (1R,2R)-(+)-dihydroxytetrahydronaphthalene in homogenous citrate buffer/ionic liquid mixtures, using t-butyl hydroperoxide as O2 donor. It tolerates up to 30 (v/v) 1,3-dimethylimidazolium methylsulfate or 1-butyl-3-methylimidazolium methylsulfate. The enzyme activity in these ionic liquid co-solvent systems is retained for 24 h, but it falls to 3 h using non-ionic organic solvents such as t-BuOH or acetone.
Sigma-Aldrich
1,2-Dihydronaphthalene, 95%