Merck
  • Presence of prophenoloxidase in the humoral fluid of amphioxus Branchiostoma belcheri tsingtauense.
Presence of prophenoloxidase in the humoral fluid of amphioxus Branchiostoma belcheri tsingtauense.
Fish & shellfish immunology (2004-08-18)
Qiuxiang Pang, Shicui Zhang, Changfa Wang, Xiaodong Shi, Yaning Sun
ABSTRACT
The presence of phenoloxidase (PO) activity in the humoral fluid of amphioxus Branchiostoma belcheri tsingtauense was electrophoretically and spectrophotometrically studied. The enzyme was present in the humoral fluid predominantly as an inactive proenzyme, prophenoloxidase (proPO). The optimum temperature for activation of the proPO ranged from 30 degrees C to 35 degrees C, and the enzyme exhibited optimum activity at pH between 7.0 and 7.5. ProPO in the humoral fluid was readily activated to active form PO by exogenous elicitors such as trypsin, zymosan and LPS. The activation of the proPO by exogenous elicitors was significantly enhanced in the presence of 10 mM Ca2+, but was susceptible to serine protease inhibitors like soybean trypsin inhibitor and p-nitrophenyl-p'-guanidinobenzoate. PAGE revealed a single band of PO activity in the humoral fluid with an apparent molecular mass of 150 kDa, which was resolved to three bands with molecular masses of 44, 46 and 72 kDa, respectively, after SDS-PAGE. This is the first report on the presence of the enzyme PO in amphioxus humoral fluid.