Merck
  • Total synthesis and structure of the ramoplanin A1 and A3 aglycons: two minor components of the ramoplanin complex.
Total synthesis and structure of the ramoplanin A1 and A3 aglycons: two minor components of the ramoplanin complex.
Proceedings of the National Academy of Sciences of the United States of America (2004-06-04)
Dongwoo Shin, Yosup Rew, Dale L Boger
ABSTRACT
Ramoplanin is a potent antibiotic, first disclosed in 1984, that acts by inhibiting bacterial cell-wall biosynthesis. The original ramoplanin complex was shown to consist of a mixture of three closely related compounds, ramoplanin A1-A3, of which ramoplanin A2 is the most abundant. The structure of ramoplanin A2 was unambiguously established first through a series of extensive spectroscopic studies, allowing complete stereochemical assignments and subsequently providing a minor reassignment of the side-chain double-bond stereochemistry and, most recently, through total synthesis of authentic material. Here we report the total syntheses of the aglycons of the minor components of the ramoplanin complex, A1 and A3, which unambiguously establish their structure and provide an expected structural revision for the lipid side-chain double-bond stereochemistry.