Merck
  • Activation of c-Jun N-terminal kinase in A549 lung carcinoma cells by sodium dichromate: role of dissociation of apoptosis signal regulating kinase-1 from its physiological inhibitor thioredoxin.
Activation of c-Jun N-terminal kinase in A549 lung carcinoma cells by sodium dichromate: role of dissociation of apoptosis signal regulating kinase-1 from its physiological inhibitor thioredoxin.
Toxicology (2004-03-09)
N J Hodges, D Smart, A J Lee, N A Lewis, J K Chipman
ABSTRACT
Changes in the components of the Jun N-terminal kinase (JNK) signalling pathway were investigated in human A549 lung carcinoma cells treated with sodium dichromate. Sodium dichromate (100 microM, 0-6h) failed to activate nuclear factor kappa B (NF-kappaB) as determined by a lack of nuclear translocation of p65 but resulted in Jun N-terminal kinase activation as assessed by phospho-Jun N-terminal kinase Western blotting in a dose-dependent (>25 microM) and time-dependent (>1h) manner. In addition, c-Jun, a downstream target of Jun N-terminal kinase signalling was also activated with a similar dose- and time-dependency at the level of both protein expression and degree of phosphorylation. In contrast, sodium dichromate treatment had no effect on levels of phospho-p38. Immunoprecipitation demonstrated that apoptosis signal regulating kinase-1 (ASK-1), an upstream activator of Jun N-terminal kinase was dissociated from its inhibitory partner thioredoxin (Trx) in response to sodium dichromate (100 microM, 4h) treatment. This treatment was also associated with a transient (2h) increase in cytosolic levels of thioredoxin but no nuclear translocation of thioredoxin was observed. In conclusion, sodium dichromate had a stimulatory effect on the Jun N-terminal kinase signalling pathway in A549 cells, resulting in activation of downstream effector molecules. We hypothesise that dissociation of apoptosis signal regulating kinase-1 from thioredoxin may be at least partially responsible for Jun N-terminal kinase activation.
Sigma-Aldrich
Sodium dichromate dihydrate, AR, ≥99.5%
Sigma-Aldrich
Sodium dichromate dihydrate, LR, ≥99%
Sigma-Aldrich
Sodium dichromate dihydrate, ACS reagent, ≥99.5%