Merck
A new technique for staining mast cells using ferroin.
Biotechnic & histochemistry : official publication of the Biological Stain Commission (2004-03-03)
V H Tomasi, S C Orrea, A R Raimondi, M E Itoiz
ABSTRACT
We describe here a new method for specific staining of mast cells using ferroin. Different hamster tissues were fixed in 4% formalin and processed for paraffin embedding. Sections were stained with hematoxylin followed by ferroin acidified with 2.5 N sulfuric acid to pH 4.0. Mast cells stained an intense orange color that contrasted markedly with bluish violet nuclei. High contrast was also observed when ferroin colored sections were counterstained with light green instead of hematoxylin. To evaluate the specificity of the stain, hamster cheek pouch sections were stained with toluidine blue, alcian blue-safranin O, and ferroin. Quantitative evaluation of mast cells stained with the three techniques showed no statistical difference. The simplicity and selectivity of this method is sufficient for image analysis of mast cells.
Sigma-Aldrich
Ferroin indicator solution, 0.1 wt. % in H2O
Sigma-Aldrich
Ferroin indicator solution, 2 % (w/v)