Merck
Photochemical reaction monitoring by ultra-violet spectrophotometry.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2002-12-13)
B Roig, E Touraud, O Thomas
ABSTRACT
Within the framework of the monitoring of the trichloroacetylchloride (TCAC) photosynthesis, ultra-violet (UV) spectrophotometry is proposed as a simple and rapid tool allowing, in real time, the control of the process efficiency. A good correlation has been obtained between the results acquired by this alternative method and the standard gas chromatography.
Product Description
Supelco
Trichloroacetyl chloride, for GC derivatization, ≥98.5%
Sigma-Aldrich
Trichloroacetyl chloride, 99%