- [Studies on chemical constituents of Curcuma aromatica salisb].
[Studies on chemical constituents of Curcuma aromatica salisb].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2002-09-06)
K X Huang, Z M Tao, A J Zhang, S L Peng, L S Ding
PMID12212100
ABSTRACT
To study the effective ingredients of Curcuma aromatica. Solvent extraction was used. The constituents were isolated with resin D-101 silica gel column and thin-layer chromatography, and the structures were elucidated by physico-chemical properties and spectral analysis. Curdione, neocurdione, curcumol, tetramethylpyrazine and (R)-(+)-1,2-hexadecanediol were isolated from C. aromatica. Neocurdione and (R)-(+)-1,2-hexadecanediol were isolated from C. aromatica for the first time, and was isolated from Curcuma and reported for the first time.