Merck
  • In situ localization of endogenous cytokinins during shooty tumor development on Eucalyptus globulus Labill.
In situ localization of endogenous cytokinins during shooty tumor development on Eucalyptus globulus Labill.
Planta (2001-08-29)
A Azmi, W Dewitte, H Van Onckelen, D Chriqui
ABSTRACT
Our previous results demonstrated that endogenous cytokinins are involved in the shooty potential of tumors initiated on Eucalyptus globulus plantlets inoculated with Agrobacterium tumefaciens strain 82.139 [A. Azmi et al. (1997a) Plant Sci 127: 81-90]. In order to investigate whether or not these hormones are distributed homogeneously in the tumors prior to the onset of bud regeneration, decapitated hypocotyls were inoculated with the strain C58pMP90/T139 GUS-INT harboring the wild transferred DNA (T-DNA) of strain 82.139 tagged with the beta-glucuronidase (gus)-reporter gene. In situ immunolocalization of zeatin, dihydrozeatin and isopentenyladenine was performed in the developing tumors and combined with the histo-enzymological beta-glucuronidase assay. It was found that the expression of the T-DNA was restricted to only some small areas located deeply in the tumors. These sites were also provided with a high cytokinin signal while the untransformed parts of the tumors displayed a weaker signal, except in the early differentiating tracheary elements. The regenerated buds were untransformed and originated from superficial parts of the tumors provided with a moderate signal for cytokinins. The method of colocalization of both cytokinins and gus expression developed here might be helpful for further studies concerning the role of these hormones in controlling gene expression at cell and tissue levels.
Sigma-Aldrich
6-(γ,γ-Dimethylallylamino)purine, BioReagent, suitable for plant cell culture, 1 mg/mL
Sigma-Aldrich
6-(γ,γ-Dimethylallylamino)purine, suitable for plant cell culture, BioReagent, ≥98.5%
Sigma-Aldrich
6-(γ,γ-Dimethylallylamino)purine, suitable for plant cell culture, BioReagent, ≥90%
Sigma-Aldrich
6-(γ,γ-Dimethylallylamino)purine, Vetec, reagent grade, ≥98.5%