Merck
  • Structural analysis of the catalytic and binding sites of Clostridium botulinum neurotoxin B.
Structural analysis of the catalytic and binding sites of Clostridium botulinum neurotoxin B.
Nature structural biology (2000-08-10)
S Swaminathan, S Eswaramoorthy
ABSTRACT
Clostridium botulinum neurotoxins are among the most potent toxins to humans. The crystal structures of intact C. botulinum neurotoxin type B (BoNT/B) and its complex with sialyllactose, determined at 1. 8 and 2.6 A resolution, respectively, provide insight into its catalytic and binding sites. The position of the belt region in BoNT/B is different from that in BoNT/A; this observation presents interesting possibilities for designing specific inhibitors that could be used to block the activity of this neurotoxin. The structures of BoNT/B and its complex with sialyllactose provide a detailed description of the active site and a model for interactions between the toxin and its cell surface receptor. The latter may provide valuable information for recombinant vaccine development.
Sigma-Aldrich
3′-Sialyllactose, from bovine milk or colostrum, ≥97% (HPLC)