Merck
  • Inhibition of mitogen-activated protein kinase kinase blocks T cell proliferation but does not induce or prevent anergy.
Inhibition of mitogen-activated protein kinase kinase blocks T cell proliferation but does not induce or prevent anergy.
Journal of immunology (Baltimore, Md. : 1950) (1998-05-09)
D R DeSilva, E A Jones, M F Favata, B D Jaffee, R L Magolda, J M Trzaskos, P A Scherle
ABSTRACT
Three mitogen-activated protein kinase pathways are up-regulated during the activation of T lymphocytes, the extracellular signal-regulated kinase (ERK), Jun NH2-terminal kinase, and p38 mitogen-activated protein kinase pathways. To examine the effects of blocking the ERK pathway on T cell activation, we used the inhibitor U0126, which has been shown to specifically block mitogen-activated protein kinase/ERK kinase (MEK), the kinase upstream of ERK. This compound inhibited T cell proliferation in response to antigenic stimulation or cross-linked anti-CD3 plus anti-CD28 Abs, but had no effect on IL-2-induced proliferation. The block in T cell proliferation was mediated by down-regulating IL-2 mRNA levels. Blocking Ag-induced proliferation by inhibiting MEK did not induce anergy, unlike treatments that block entry into the cell cycle following antigenic stimulation. Surprisingly, induction of anergy in T cells exposed to TCR cross-linking in the absence of costimulation was also not affected by blocking MEK, unlike cyclosporin A treatment that blocks anergy induction. These results suggest that inhibition of MEK prevents T cell proliferation in the short term, but does not cause any long-term effects on either T cell activation or induction of anergy. These findings may help determine the viability of using mitogen-activated protein kinase inhibitors as immune suppressants.
Sigma-Aldrich
U0126 ethanolate, ≥98% (HPLC), powder