Merck
  • Protein chemical characterization of Mucor pusillus aspartic proteinase. Amino acid sequence homology with the other aspartic proteinases, disulfide bond arrangement and site of carbohydrate attachment.
Protein chemical characterization of Mucor pusillus aspartic proteinase. Amino acid sequence homology with the other aspartic proteinases, disulfide bond arrangement and site of carbohydrate attachment.
FEBS letters (1988-08-01)
M Baudys, S Foundling, M Pavlík, T Blundell, V Kostka
ABSTRACT
The amino acid sequence of Mucor pusillus aspartic proteinase was determined by analysis of fragments obtained from cleavage of the enzyme by CNBr and limited tryptic digestion. The proteinase is a single polypeptide chain protein containing 361 amino acid residues, cross-linked by two disulfide bonds. A sugar moiety composed of two GlcNAc residues and four neutral sugar residues is asparagine-linked to the chain. The sequence of M. pusillus proteinase is highly homologous with the M. miehei proteinase (83% identity). The homology with other aspartic proteinases is low (22-24%) and indicates that the Mucor proteinases diverged at an early evolutionary phase. The most conservative regions of the molecule are those involved in catalysis and forming the binding cleft and the core region of the molecule.
Sigma-Aldrich
Rennin from Mucor miehei, lyophilized, powder, slightly brown, ~0.1 U/mg