Merck
  • Chemo-enzymatic conversion of glucose into 5-hydroxymethylfurfural in seawater.
Chemo-enzymatic conversion of glucose into 5-hydroxymethylfurfural in seawater.
ChemSusChem (2012-05-25)
Philipp M Grande, Christian Bergs, Pablo Domínguez de María
ABSTRACT
Do you sea water? Water consumption will be a challenge in biorefineries, and the use of non-drinkable sources of water will be preferred. Herein, glucose is converted into 5-hydroxymethylfurfural (HMF) in a chemo-enzymatic one-pot, two-step procedure, involving immobilized glucose isomerase to produce fructose and oxalic acid to dehydrate it to HMF.
Sigma-Aldrich
Glucose Isomerase from Streptomyces murinus, ≥350 U/g