Merck
  • Alkadienyl and alkenyl itaconic acids (ceriporic acids G and H) from the selective white-rot fungus Ceriporiopsis subvermispora: a new class of metabolites initiating ligninolytic lipid peroxidation.
Alkadienyl and alkenyl itaconic acids (ceriporic acids G and H) from the selective white-rot fungus Ceriporiopsis subvermispora: a new class of metabolites initiating ligninolytic lipid peroxidation.
Organic & biomolecular chemistry (2012-06-29)
Hiroshi Nishimura, Midori Sasaki, Hirofumi Seike, Masaharu Nakamura, Takashi Watanabe
ABSTRACT
New ceriporic acids-alkadienyl and alkenyl itaconic acids having a bis-allyl (3-[(Z,Z)-hexadec-7,10-dienyl]-itaconic acid; ceriporic acid G) and a monoene (3-[(Z)-octadec-9-enyl]-itaconic acid; ceriporic acid H) structure in their side chains-were isolated from the cultures of the selective lignin-degrading fungus Ceriporiopsis subvermispora. The new metabolites ceriporic acid G and H were synthesized by a cross-aldol condensation and a Grignard substitution reaction, respectively. Ceriporic acid G triggered the manganese peroxidase (MnP)-catalyzed lipid peroxidation and decomposed a recalcitrant non-phenolic lignin substructure model compound. Except for simple fatty acids, this is the first report of a fungal metabolite that induced ligninolytic lipid peroxidation.