- [The effect of acute alcohol intoxication on some parameters of nitrogenous metabolism in rats with alloxan diabetes].
[The effect of acute alcohol intoxication on some parameters of nitrogenous metabolism in rats with alloxan diabetes].
Biomeditsinskaia khimiia (2012-06-26)
N V Zharkova, P P Potapov, A Iu Stel'makh
PMID22724361
ABSTRACT
Acute alcohol intoxication in rats with alloxan diabetes is accompanied by the increase of urea and uric acid and by the decrease in free fatty acids in serum. In the liver of experimental animals the increase of activity of glutamate dehydrogenase, AMP deaminase, and tyrosine transaminase was found.
Sigma-Aldrich
Glutaminase from Escherichia coli, Grade VII, lyophilized powder, 500-1,500 units/mg protein