Merck
  • Characterization of dextran-producing Weissella strains isolated from sourdoughs and evidence of constitutive dextransucrase expression.
Characterization of dextran-producing Weissella strains isolated from sourdoughs and evidence of constitutive dextransucrase expression.
FEMS microbiology letters (2010-08-21)
Marie-Sophie Bounaix, Hervé Robert, Valérie Gabriel, Sandrine Morel, Magali Remaud-Siméon, Bruno Gabriel, Catherine Fontagné-Faucher
ABSTRACT
The study of exopolysaccharide production by heterofermentative sourdough lactic acid bacteria has shown that Weissella strains isolated from sourdoughs produce linear dextrans containing α-(1→6) glucose residues with few α-(1→3) linkages from sucrose. In this study, several dextran-producing strains, Weissella cibaria and Weissella confusa, isolated from sourdough, were characterized according to carbohydrate fermentation, repetitive element-PCR fingerprinting using (GTG)(5) primers and glucansucrase activity (soluble or cell-associated). This study reports, for the first time, the characterization of dextransucrase from Weissella strains using sodium dodecyl sulfate-polyacrylamide gel electrophoresis and in situ polymer production (after incubation with sucrose) from enzymatic fractions harvested from both sucrose and glucose culture media. Results demonstrate that dextransucrase activity was mainly soluble and associated with a constitutive 180-kDa protein. In addition, microsequencing of the active dextransucrase from W. cibaria LBAE-K39 allowed the design of specific primers that could detect the presence of glucansucrase encoding genes similar to GTFKg3 of Lactobacillus fermentum Kg3 and to DSRWC of W. cibaria CMU. This study hence indicates that sourdough Weissella strains synthesize original dextransucrase.