Merck
  • Cephalosporolides H and I, Two Novel Lactones from a Marine-Derived Fungus, Penicillium sp.
Cephalosporolides H and I, Two Novel Lactones from a Marine-Derived Fungus, Penicillium sp.
Archives of pharmacal research (2007-08-21)
Xiang Li, Yanhua Yao, Yinan Zheng, Isabel Sattler, Wenhan Lin
ABSTRACT
Two novel lactones, Cephalosporolides H (1) and 1 (2), were isolated from the lyophilized culture broth of the marine-derived fungus, Penicillium sp. The structures were elucidated on the basis of extensive 1D- and 2D-NMR, as well as HRESI-MS spectroscopic analyses. The relative stereochemistries of the compounds were assessed by comparison of the NOESY analysis and spectral data with those in the literature.