Merck
  • Structurally distinct active sites in the copper(II)-substituted aminopeptidases from Aeromonas proteolytica and Escherichia coli.
Structurally distinct active sites in the copper(II)-substituted aminopeptidases from Aeromonas proteolytica and Escherichia coli.
Journal of the American Chemical Society (2002-10-31)
Brian Bennett, William E Antholine, Ventris M D'souza, Guanjing Chen, Leila Ustinyuk, Richard C Holz
ABSTRACT
The aminopeptidase from Aeromonas proteolytica (AAP) was titrated with copper, which bound sequentially at two distinct sites. Both the mono- and disubstituted forms of AAP exhibited catalytic hyperactivity relative to the native dizinc enzyme. Monosubstituted AAP exhibited an axial Cu(II) EPR spectrum with slight pH dependence: at pH 6.0 g(parallel) = 2.249, g( perpendicular ) = 2.055, and A(parallel)((63/65)Cu) = 1.77 x 10(-)(2) cm(-)(1), whereas at pH 9.65 g(parallel) = 2.245, g( perpendicular ) = 2.056, and A(parallel)((63/65)Cu) = 1.77 x 10(-)(2) cm(-)(1). These data indicate oxygen and nitrogen ligation of Cu. AAP further substituted with copper exhibited a complex signal with features around g approximately 2 and 4. The features at g approximately 4 were relatively weak in the B(0) perpendicular B(1) (perpendicular) mode EPR spectrum but were intense in the B(0) parallel B(1) (parallel) mode spectrum. The g approximately 2 region of the perpendicular mode spectrum exhibited two components, one corresponding to mononuclear Cu(II) with g(parallel) = 2.218, g( perpendicular ) = 2.023, and A(parallel)((63/65)Cu) = 1.55 x 10(-)(2) cm(-)(1) and likely due to adventitious binding of Cu(II) to a site distant from the active site. Excellent simulations were obtained for the second component of the spectrum assuming that two Cu(II) ions experience dipolar coupling corresponding to an inter-copper distance of 5 A with the two Cu(II) g(z)() directions parallel to each other and at an angle of approximately 17 degrees to the inter-copper vector (H = betaB.g(CuA).S(CuA) + betaB.g(CuB).S(CuB) + [S.A.I](CuA) + [S.A.I](CuB) + [S(CuA).J.S(CuB)]; g(parallel(CuA,CuB)) = 2.218, g( perpendicular )((CuA,CuB)) = 2.060; A(parallel(CuA,CuB))((63/65)Cu) = 1.59 x 10(-)(2) cm(-)(1), J(isotropic) = 50 cm(-)(1), r(Cu)(-)(Cu) = 4.93 A, and chi = 17 degrees ). The exchange coupling between the two copper ions was found to be ferromagnetic as the signals exhibited Curie law temperature dependence. The Cu-Cu distance of approximately 5 A indicated by EPR was significantly higher than the inter-zinc distance of 3.5 A in the native enzyme, and the dicopper species therefore represents a novel dinuclear site capable of catalysis of hydrolysis. In contrast to AAP, the related methionyl aminopeptidase from Escherichia coli (EcMetAP) was found to bind only one Cu(II) ion despite possessing a dinuclear binding site motif. A further difference was the marked pH dependence of the signal in EcMetAP, suggestive of a change in ligation. The structural motifs of these two Cu(II)-substituted aminopeptidases provide important insight into the observed catalytic activity.
Sigma-Aldrich
Aminopeptidase His-tagged from Vibrio proteolyticus