Merck
In vitro fluorescent analysis of preprotein import into chloroplasts.
Bioscience, biotechnology, and biochemistry (2011-10-08)
Sattasuk Kwanchanok, Hitoshi Inoue, Mitsuru Akita
ABSTRACT
Despite recent progress in fluorescence techniques employed to observe protein localization in living cells, the in vitro chloroplastic protein transport assay remains a useful tool for determining the destinations of proteins. Although an in vitro synthesized, radiolabeled precursor protein is frequently used as the transport substrate, we have developed a transport assay system with a non-radiolabeled precursor protein that carries an epitope tag and is overexpressed in Escherichia coli. Thus, a transported protein can be detected by immunoblotting (Inoue et al., Plant Physiol. Biochem., 46, 541-549 (2008)). Here, we propose another in vitro protein transport system that combines fluorescence techniques. We attempted to use two types of precursors: a green fluorescent protein (GFP)-fused precursor and a fluorescent dye-labeled one. Both were successfully imported into chloroplasts. However, the fluorescent dye-labeled precursor was more advantageous than the GFP-fused precursor in the in vitro system.
Sigma-Aldrich
N-(5-Fluoresceinyl)maleimide, suitable for fluorescence, ≥90% (HPLC), BioReagent