Merck
  • Transformation of 5 alpha-cholest-7-en-3 beta-ol to cholesterol and cholestanol in cerebrotendinous xanthomatosis.
Transformation of 5 alpha-cholest-7-en-3 beta-ol to cholesterol and cholestanol in cerebrotendinous xanthomatosis.
Journal of lipid research (1974-05-01)
G S Tint, G Salen
ABSTRACT
The metabolism of Delta(7)-cholestenol, cholesterol, and cholestanol was examined in a patient with cerebrotendinous xanthomatosis after intravenous pulse-labeling with a mixture of dl-[2-(14)C]mevalonate and stereospecific 3S,4S,3R,4R-[4-(3)H]mevalonate. Silver nitrate and reversed-phase thin-layer chromatography were used to purify the sterols isolated from the feces, and their identities were confirmed by gas-liquid chromatography-mass spectrometry. The specific activities were determined and plotted as a function of time. Isotope ratio measurements and specific activity decay curves showed that sterol synthesis proceeded in the following sequence: mevalonate, squalene, lanosterol, Delta(7)-cholestenol, cholesterol, cholestanol. Labeled cholesterol precursors might be advantageously used to measure changes in cholesterol synthesis because they appear to equilibrate rapidly and have very short turnover times.