Merck
  • ADAMTS4 is a crucial proteolytic enzyme for versican cleavage in the amnion at parturition.
ADAMTS4 is a crucial proteolytic enzyme for versican cleavage in the amnion at parturition.
Communications biology (2024-03-10)
Meng-Die Li, Jiang-Wen Lu, Fan Zhang, Wen-Jia Lei, Fan Pan, Yi-Kai Lin, Li-Jun Ling, Leslie Myatt, Wang-Sheng Wang, Kang Sun
ABSTRACT
Hyalectan cleavage may play an important role in extracellular matrix remodeling. However, the proteolytic enzyme responsible for hyalectan degradation for fetal membrane rupture at parturition remains unknown. Here, we reveal that versican (VCAN) is the major hyalectan in the amnion, where its cleavage increases at parturition with spontaneous rupture of membrane. We further reveal that ADAMTS4 is a crucial proteolytic enzyme for VCAN cleavage in the amnion. Inflammatory factors may enhance VCAN cleavage by inducing ADAMTS4 expression and inhibiting ADAMTS4 endocytosis in amnion fibroblasts. In turn, versikine, the VCAN cleavage product, induces inflammatory factors in amnion fibroblasts, thereby forming a feedforward loop between inflammation and VCAN degradation. Mouse studies show that intra-amniotic injection of ADAMTS4 induces preterm birth along with increased VCAN degradation and proinflammatory factors abundance in the fetal membranes. Conclusively, there is enhanced VCAN cleavage by ADAMTS4 in the amnion at parturition, which can be reenforced by inflammation.