Merck
  • Decreased expression of p53 is associated with down expression of zyxin in breast cancer.
Decreased expression of p53 is associated with down expression of zyxin in breast cancer.
Health science reports (2024-08-05)
Rezvan Rostampour, Kiana Bahremand, Hossein Mohammadi, Seyed Askar Roghani, Ebrahim Shakiba, Mohammad Taghi Goodarzi, Soheila Asadi
ABSTRACT
Breast cancer (BC) is considered one of the most common malignant tumors leading to death in women, and genetic factors have a crucial role in BC pathogenesis. Zyxin (ZYX) is one of these factors that may be important in p53 level and function. Thus, the present work aimed to investigate the ZYX gene and protein expression in tumor tissue and matched margin tissue and its correlation with the p53 expression. In a present case-control study, 30 tumors and 30 matched margin tissues were obtained from Iran Tumor Bank/Tehran University of Medical Sciences. Real-time polymerase chain reaction and western blot analysis techniques were applied to evaluate the genes and protein expression, respectively. The data showed that expression of the ZYX gene in tumor tissues significantly decreased (p = 0.0274) compared to matched margin tissues. In contrast, the p53 gene expression in tumor tissues had no significant difference with matched margin tissues. Additionally, we observed that ZYX and p53 genes expression in tumor tissues of estrogen receptor-positive patients had significant elevation than estrogen receptor-negative patients (p < 0.001, p < 0.001, respectively). The data of the western blot analysis technique showed that protein expression of ZYX (p = 0.0024) and P53 protein (p = 0.0218) in tumor tissues was significantly reduced compared to matched margin tissues. Additionally, our analysis showed a direct and significant correlation between the expression of ZYX and p53 proteins (r = 0.7797, p = 0.0126) and expression of ZYX and p53 genes (r = 0.3079, p = 0.0187). Based on our observation, ZYX might have a tumor suppressor role and is associated with p53.