Merck
  • Identification of the function of FOSB in cholangiocarcinoma using bioinformatics analysis.
Identification of the function of FOSB in cholangiocarcinoma using bioinformatics analysis.
Translational cancer research (2024-01-09)
Yihang Zhao, Hong Tang, Yaxian Kuai, Jianhua Xu, Bin Sun, Yang Li
ABSTRACT
Exploring the potential mechanism of cholangiocarcinoma (CCA) metabolic reprogramming is significant for guiding clinical treatment. However, related research and exploration are still lacking. Therefore, we aimed to identify a reliable metabolism-related gene or biomarker of CCA using bioinformatics analysis. The GSE26566, GSE45001, and GSE132305 datasets were obtained from the Gene Expression Omnibus (GEO) database. Differently expressed genes (DEGs) between CCA tissues and adjacent tissues were screened out. The key gene was identified through enrichment and functional analysis, and its immune and clinical correlation was investigated utilizing the Tumor Immune Evaluation Resource (TIMER2.0), the Tumor-Immune System Interactions Database (TISIDB), the Gene Expression Profiling Interactive Analysis (GEPIA2), and the Kaplan-Meier Plotter. Finally, immunohistochemistry (IHC) was performed to validate the results. By analysis, the expression of FBJ murine osteosarcoma viral oncogene homolog B (FOSB) was significantly downregulated in CCA tissues when compared with adjacent tissues. Moreover, the expression levels of FOSB positively correlated with tumor-infiltrating immune cells in most tumors, and patients with high FOSB expression tended to have a better prognosis. The FOSB and SIRT3/HIF1A axes have similar expression trends and metabolic functions in CCA cells, and the correlation between of them was preliminarily explored by IHC experiments. The expression levels of FOSB are closely related to the prognosis of CCA patients, which may be a predictive indicator for prognosis and immunotherapy.